Aluminiumkaliumsulfat Dodecahydrat 98,0-102,0% (durch EDTA Titration), Kristalle, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor

Synonyme: Alumen, Kalialaun, Aluminiumkaliumbis(sulfat) Dodecahydrat

0546-01EA 247.83 EUR
0546-01 0546-20
Aluminiumkaliumsulfat Dodecahydrat 98,0-102,0% (durch EDTA Titration), Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Aluminiumkaliumsulfat Dodecahydrat

Crystals. Lot analysis on label.

Formel: AlK(SO₄)₂·12H₂O
Schmelzpunkt: 92,5 °C
Dichte: 1,72 g/cm³ (20 °C)
MDL: MFCD00149143
CAS-Nummer: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 98.0 - 102.0%
For Laboratory,Research,or Manufacturing Use 99.0 - 100.5%
Assay (AlK(SO₄)₂·12H₂O) (by EDTA titrn) 98.0 - 102.0 %
Assay (dried basis) 99.0 - 100.5 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 2.0 - 4.0
Ammonium (NH₄) ≤0.005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Sodium (Na) ≤0.02 %
Chloride (Cl) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR