L(+)-Ascorbinsäure 99,9998 Metals basis, TraceSELECT™, Fluka™
Lieferant: Honeywell Chemicals
Synonyme:
L-Threoascorbinsäure, Acidum ascorbicum, Vitamin C
05878-100G
05878-100GEA
158.05
EUR
05878-100G
L(+)-Ascorbinsäure 99,9998 Metals basis, TraceSELECT™, Fluka™
L(+)-Ascorbinsäure
TraceSELECT™ Ultra and TraceSELECT
- Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Formel:
C₆H₈O₆ Molecular Weight: 176.13 g/mol Siedepunkt: 553 Schmelzpunkt: 190…194 °C Dichte: 1.65 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064328 CAS-Nummer: 50-81-7 EINECS: 200-066-2 REACH: exempt Merck Index: 13,00837 |
|
Spezifikation Testergebnisse
Assay | Min. 99.0 % |
Specific rotation (a) 20/D; c=10 in H₂O | 20.5 - 21.5 ° |
Silver (Ag) | Max. 0.01 ppm |
Aluminium (Al) | Max. 0.05 ppm |
Arsenic (As) | Max. 0.005 ppm |
Gold (Au) | Max. 0.01 ppm |
Barium (Ba) | Max. 0.01 ppm |
Beryllium (Be) | Max. 0.01 ppm |
Bismuth (Bi) | Max. 0.01 ppm |
Calcium (Ca) | Max. 0.1 ppm |
Cadmium (Cd) | Max. 0.01 ppm |
Cerium (Ce) | Max. 0.01 ppm |
Cobalt (Co) | Max. 0.01 ppm |
Chromium (Cr) | Max. 0.05 ppm |
Cesium (Cs) | Max. 0.01 ppm |
Copper (Cu) | Max. 0.01 ppm |
Dysprosium (Dy) | Max. 0.01 ppm |
Erbium (Er) | Max. 0.01 ppm |
Europium (Eu) | Max. 0.01 ppm |
Iron (Fe) | Max. 0.1 ppm |
Gallium (Ga) | Max. 0.01 ppm |
Gadolinium (Gd) | Max. 0.01 ppm |
Germanium (Ge) | Max. 0.05 ppm |
Hafnium (Hf) | Max. 0.01 ppm |
Mercury (Hg) | Max. 0.01 ppm |
Holmium (Ho) | Max. 0.01 ppm |
Indium (In) | Max. 0.01 ppm |
Iridium (Ir) | Max. 0.01 ppm |
Potassium (K) | Max. 0.1 ppm |
Lanthanum (La) | Max. 0.01 ppm |
Lithium (Li) | Max. 0.01 ppm |
Lutetium (Lu) | Max. 0.01 ppm |
Magnesium (Mg) | Max. 0.05 ppm |
Manganese (Mn) | Max. 0.01 ppm |
Molybdenum (Mo) | Max. 0.01 ppm |
Sodium (Na) | Max. 2 ppm |
Niobium (nb) | Max. 0.01 ppm |
Neodymium (nd) | Max. 0.01 ppm |
Nickel (Ni) | Max. 0.01 ppm |
Lead (Pb) | Max. 0.01 ppm |
Palladium (Pd) | Max. 0.01 ppm |
Praseodymium (Pr) | Max. 0.01 ppm |
Platinum (Pt) | Max. 0.01 ppm |
Rubidium (Rb) | Max. 0.01 ppm |
Rhenium (Re) | Max. 0.01 ppm |
Rhodium (Rh) | Max. 0.01 ppm |
Ruthenium (Ru) | Max. 0.01 ppm |
Antimony (Sb) | Max. 0.01 ppm |
Scandium (Sc) | Max. 0.05 ppm |
Selenium (Se) | Max. 0.01 ppm |
Samarium (Sm) | Max. 0.01 ppm |
Tin (Sn) | Max. 0.01 ppm |
Strontium (Sr) | Max. 0.01 ppm |
Tantalum (Ta) | Max. 0.01 ppm |
Terbium (Tb) | Max. 0.01 ppm |
Tellurium (Te) | Max. 0.01 ppm |
Thorium (Th) | Max. 0.01 ppm |
Titanium (Ti) | Max. 0.05 ppm |
Thallium (Tl) | Max. 0.01 ppm |
Thulium (Tm) | Max. 0.01 ppm |
Uran (U) | Max. 0.01 ppm |
Vanadium (V) | Max. 0.01 ppm |
Tungsten (W) | Max. 0.01 ppm |
Yttrium (Y) | Max. 0.01 ppm |
Ytterbium (Yb) | Max. 0.01 ppm |
Zinc (Zn) | Max. 0.05 ppm |
Zirconium (Zr) | Max. 0.01 ppm |
Chloride (Cl) | Max. 50 ppm |
Sulfate (SO₄) | Max. 20 ppm |
Ammonium (NH₄) | Max. 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...