Ammoniumacetat für die HPLC, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Essigsäure Ammoniumsalz

0599-08EA 307.16 EUR
0599-08
Ammoniumacetat für die HPLC, J.T.Baker®
Ammoniumacetat
Formel: H₃CCOONH₄
Molecular Weight: 77,08 g/mol
Schmelzpunkt: 114 °C
Dichte: 1,17 g/cm³ (20 °C)
Flammpunkt: 136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013066
CAS-Nummer: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use = 97 %
Assay (CH₃COONH₄) (by acid-base titration) ≥97 %
Insoluble Matter ≤0.005 %
Residue after Ignition ≤0.010 %
pH of 5% Solution at 25°C 6.7 - 7.3
Nitrate (NO₃) ≤0.001 %
Sulfate (SO₄) ≤0.001 %
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 350 nm ≤0.010
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 280 nm ≤0.010
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 254 nm ≤0.020
Chloride (Cl) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR