Phenol 99,0-100,5% (durch Trockensubstanz), Kristalle USP, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

0605-04EA 263.14 EUR
0605-04 0605-06
Phenol 99,0-100,5% (durch Trockensubstanz), Kristalle USP, Macron Fine Chemicals™
Formel: C₆H₅OH
Siedepunkt: 181,8 °C (1013 hPa)
Dichte: 1,06 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002143
CAS-Nummer: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1,II

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product 99.0 - 100.5%
Meets U.S.P Requirements ≤2.0 ppm
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical Passes Test
USP - Assay (C₆H₅OH) (anhydrous basis) 99.0 - 100.5 %
USP - Appearance of Melted Sample Passes Test
USP - Benzene ≤2.0 ppm
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Clarity of Solution and Reaction Passes Test
USP - Congealing Temperature ≥39 °C
USP - Nonvolatile Residue ≤0.05 %
USP - Water (H₂O)(by Karl Fischer titrn) ≤0.5 %
Appearance (colorless to light pink, crystalline mass)
No Class 2 or 3 solvents are expected to be present. The Class 1
solvent Benzene is used as a raw material in a precursor to phenol. The product is tested for any presence of benzene and the result
reported on the Certificate of Analysis.
Product contains about 20 ppm Citric Acid as a preservative.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR