Phenol USP, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: Phenylalkohol, Hydroxybenzol, Carbolsäure

0610-06EA 1037.64 EUR
0610-06
Phenol USP, Macron Fine Chemicals™
Phenol
Formel: C₆H₅OH
Molecular Weight: 94.11 g/mol
Siedepunkt: 181.7
Schmelzpunkt: 40.8
Flammpunkt: 81
MDL: MFCD00002143
CAS-Nummer: 108-95-2
UN: 1671
ADR: 6.1 II
Merck Index: 12,07390

Bestellen

Spezifikation Testergebnisse

Meets U.S.P Requirements
GMP Manufactured Product
Meets J.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical = 89.0 %
USP - Assay (C₆H₅OH) ≥89.0 %
USP - Benzene ≤2.0 ppm
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Clarity of Solution and Reaction Passes Test
USP - Nonvolatile Residue ≤0.05 %
Distilling Range - Initial Temperature (For Information Only) = 182.5 °C
Distilling Range - Final Temperature ≤182.5 °C
Stabilizer Passes Test
JP - Identification (1) Passes Test
JP - Identification (2) Passes Test
JP - Boiling Point ≤182 °C
JP - Clarity and Color of Solution and Acidity or Alkalinity Passes Test
JP - Residue on Evaporation ≤0.05 %
JP - Assay ≥88.0 %
Appearance (colorless to slightly pink liquid)
No Class 2 or 3 solvents are expected to be present. The Class 1
solvent Benzene is used as a raw material in a precursor to phenol. The product is tested for any presence of benzene and the result
reported on the Certificate of Analysis.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR