Dodecyltrimethylammoniumchlorid 50%, hochrein

Lieferant: VWR Chemicals
Achtung

Synonyme: (1-Dodecyl)trimethylammonium chloride, N,N,N-Trimethyldodecan-1-aminium chloride, DTAC

0624-500MLEA 281 EUR
0624-500ML
Dodecyltrimethylammoniumchlorid 50%, hochrein
Dodecyltrimethylammonium chloride

Cationic - positively-charged (quaternary ammonium compounds). Utilised for applications ranging from solubilizing membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis.

Formel: C₁₅H₃₄ClN
Molecular Weight: 263,89 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00041974
CAS-Nummer: 112-00-5
EINECS: 203-927-0

Bestellen

Spezifikation Testergebnisse

Activity ( conc.) 49,0 - 52,0 %
Free Amine & Amine HCl < 2,0 %
pH (10%, Water) at 25 °C 6,0 - 9,0
Purity (C₁₅H₃₄ClN) ≥98,5%
Specific Gravity Report value

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR