3-Amino-9-ethylcarbazol (AEC) ≥97,0% (durch HPLC), LiChropur™ Derivatisierungsreagenz, für die HPLC, Supelco®

Lieferant: Merck
Gefahr

Synonyme: 9-Ethylcarbazol-3-ylamin, AEC

06696-1G
06696-1GEA 28.1 EUR
06696-1G
3-Amino-9-ethylcarbazol (AEC) ≥97,0% (durch HPLC), LiChropur™ Derivatisierungsreagenz, für die HPLC, Supelco®
3-Amino-9-ethyl carbazole (AEC)

3-Amino-9-ethylcarbazole is a peroxidase substrate suitable for use in immunoblotting and immunohistochemical staining procedures. This substrate produces an insoluble end product that is red in color and can be observed visually.

Formel: C₁₄H₁₄N₂
Molecular Weight: 210,28 g/mol
MDL: MFCD00004964
CAS-Nummer: 132-32-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b91b6dc6867b4a2947566c143f622e5d23713ce&vid=2b91b6dc6867b4a2947566c143f622e5d23713ce&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR