4-Aminoantipyrin, Supelco®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
06800-100G
06800-25G
06800-500G
06800-100GEA
191
EUR
06800-100G
06800-25G
06800-500G
4-Aminoantipyrin, Supelco®
4-Aminoantipyrin
Formel:
C₁₁H₁₃N₃O Schmelzpunkt: 107…109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003145 CAS-Nummer: 83-07-8 EINECS: 201-452-3 Merck Index: 13,00596 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...