Supelco® Organic Reference Standard, Methyl Octanoate
Lieferant: Merck
06934-50MG
06934-50MGEA
86.4
EUR
06934-50MG
Supelco® Organic Reference Standard, Methyl Octanoate
Standards
Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.