Butylacrylat, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

06958-1ML-F
06958-1ML-FEA 35.7 EUR
06958-1ML-F
Butylacrylat, Supelco®
Butylacrylat
Formel: C₇H₁₂O₂
MDL: MFCD00008809
CAS-Nummer: 141-32-2
EINECS: 205-480-7
UN: 2348
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2a0b1839a8e6ff64ddcd5ddc1da5d412da738a3&vid=9356a8ef72aeaade23e00e74cce54fef18639b50&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR