di-Kaliumhydrogenphosphat, wasserfrei ≥98% ACS
Lieferant: VWR Chemicals
0705-2.5KGEA
454
EUR
0705-2.5KG
0705-500G
0705-12KG
0705-50KG
di-Kaliumhydrogenphosphat, wasserfrei ≥98% ACS
di-Kaliumhydrogenphosphat
Formel:
K₂HPO₄ Schmelzpunkt: 340 °C Dichte: 2,3 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011383 CAS-Nummer: 7758-11-4 EINECS: 231-834-5 Merck Index: 13,07743 |
|
Spezifikation Testergebnisse
Chloride | < 0,003% |
Heavy Metals (as Pb) | ≤ 0,0005% |
Insolubles | ≤ 0,01% |
Iron | ≤ 0,001% |
Loss on Drying | ≤ 1% |
Nitrogen Compounds | ≤ 0,001% |
pH (5%, Water) @25°C | 8,5 - 9,6 |
Purity | ≥ 98% |
Sodium | ≤ 0,05% |
Sulphate | ≤ 0,005% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...