VWR® Q Path® Freeze Gel (Glue) Mounting Medium for Cryotomy
Lieferant: VWR Q-Path Chemicals
07111245.EA
190
EUR
07111245.
VWR® Q Path® Freeze Gel (Glue) Mounting Medium for Cryotomy
Färbemittel
Mikroskopie- und Färbereagenzien
High viscosity mounting media for cryotomy. Less viscous than O.C.T.