VWR®, Pepsin 1:10000
Lieferant: VWR Chemicals
0717-25GEA
212
EUR
0717-25G
VWR®, Pepsin 1:10000
Enzyme
Pepsin is a proteolytic enzyme product derived from porcine stomach linings.
Pepsin 1:10000 is equivalent to 10000 FCC units/mg.
10000 FCC U/mg = 1,67 U/mg USP = 1,67 U/mg Ph Eur = 1,67 U/mg FIP
Loss on Drying is ≤4,0%. pH (2%, Water) @25 °C is 3,0 to 4,5.