N-Lauroylsarkosin Natriumsalz, Reagenzqualität
Lieferant: VWR Chemicals
|
Gefahr
|
Anionic - negatively-charged (carboxylates, sulfates or sulfonates in alkali metal form). Utilised for applications ranging from solubilising membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis.
- Very fine white crystalline powder
Formel:
H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa Schmelzpunkt: 45…47 °C Flammpunkt: >150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042728 CAS-Nummer: 137-16-6 EINECS: 205-281-5 UN: 2811 ADR: 6.1,II Merck Index: 12,04379 |
|
Spezifikation Testergebnisse
pH (10%, Water) @25 °C | 7.5 – 8.5 |
Purity | >92.0 % |
Sodium Laurate | ≤ 4.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...