Methylenblau Trihydrat, Reagenzqualität

Lieferant: VWR Chemicals
Achtung

Synonyme: 3,7-Bis(dimethylamino)phenothiaziniumchlorid Trihydrat, N,N,N′,N′-Tetramethylthioninchlorid Trihydrat, Methylthioniniumchlorid Trihydrat, C.I.52015

0722-100GEA 173 EUR
0722-100G
Methylenblau Trihydrat, Reagenzqualität
Methylenblau Trihydrat

Post-electrophoretic stain for DNA. Less sensitive than Ethidium bromide, but non-intercalating. Also used to stain RNA in bacteriological procedures.

Formel: C₁₆H₁₈ClN₃S·3H₂O
Molecular Weight: 373,9 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00150008
CAS-Nummer: 7220-79-3
EINECS: 200-515-2

Bestellen

Spezifikation Testergebnisse

Arsenic <0.0008%
Copper ≤0.03%
Dye Content (Dry Basis) 98 – 103%
Em (661nm, Water) ≥69000
Identification PASS/FAIL
Loss on Drying ≤15.0%
Residue on Ignition ≤1.0%
Zinc ≤0.02%
Colour Index 52015 52015

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR