Ammoniumnitrat ≥95,0%, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Ammonsalpeter
0729-05EA
246.23
EUR
0729-05
0729-01
Ammoniumnitrat ≥95,0%, BAKER ANALYZED® ACS, J.T.Baker®
Ammoniumnitrat
Formel:
NH₄NO₃ Siedepunkt: 230 °C (1013 hPa) Schmelzpunkt: 169 °C Dichte: 1,73 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011425 CAS-Nummer: 6484-52-2 EINECS: 229-347-8 UN: 1942 ADR: 5.1,III Merck Index: 13,00538 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | ≥95.0% |
For Laboratory,Research,or Manufacturing Use | ≤0.005% |
ACS - Assay (NH₄NO₃) | ≥95.0 % |
ACS - Insoluble Matter | ≤0.005 % |
ACS - Residue after Ignition | ≤0.01 % |
ACS - pH of 5% Solution at 25°C | 4.5 - 6.0 |
ACS - Sulfate (SO₄) | ≤0.002 % |
Chloride (Cl) | ≤5 ppm |
Nitrite (NO₂) | Passes Test |
Phosphate (PO₄) | ≤5 ppm |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤2.0 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...