tri-Natriumcitrat Dihydrat 99,0-100,5% (durch Trockensubstanz), Kristalle USP, FCC, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: Natriumcitrat tribasisch Dihydrat, Citronensäure Trinatriumsalz Dihydrat

0734-23EA 0 EUR
0734-23
tri-Natriumcitrat Dihydrat 99,0-100,5% (durch Trockensubstanz), Kristalle USP, FCC, Macron Fine Chemicals™
tri-Natriumcitrat Dihydrat
Formel: Na₃C₆H₅O₇·2H₂O
Molecular Weight: 294.1 g/mol
Schmelzpunkt: 150
Dichte: 1.76 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150031
CAS-Nummer: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Food GMP Manufactured Product 99.0 - 100.5%
Meets F.C.C. Requirements Passes Test
Meets U.S.P Requirements Passes Test
GMP Manufactured Product Passes Test
CAUTION: For Manufacturing, processing or repackaging 10.0 - 13.0%
Bulk Food Chemical Passes Test
Bulk Pharmaceutical Chemical Passes Test
USP - Assay (anhydrous basis) 99.0 - 100.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - Water (H₂O) 10.0 - 13.0 %
USP - Alkalinity Passes Test
USP - Tartrate Passes Test
FCC - Assay (anhydrous basis) 99.0 - 100.5 %
FCC - Identification Passes Test
FCC - Alkalinity Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Water (H₂O) 10.0 - 13.0 %
Thru U.S. No. 50 Sieve, For Information Only
Solubility (5 in 50) Passes Test
Appearance (colorless or white to slightly yellow crystals)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR