Elemental impurities - Certified Reference Materials for ICH Q3D, USP 232, USP 2232, and Ph.Eur. 5.20
Lieferant: Merck
For the new adapted USP <232> and Ph.Eur.Suppl. 9.3 chapters, Merck offers TraceCERT® element mixes with element ratio corresponding to the oral concentrations of the ICH Q3D guideline, Standard 1 is a mix which covers class 1, 2A and some of 2B elements; Standard 2 is a mix which covers the remaining 2B class elements; Standard 3 is a mix which covers all class 3 elements.
- Unique level of accuracy and lot-specific value
- Sophisticated packaging and comprehensive documentation including proper uncertainty calculation, expiry date and storage information
- Packaged in opaque and gas-tight aluminum foil bags for extended stability. Certificates are included and list up to 70 trace impurities for the TraceCERT® products.
Standards: Produced according to ISO 17034 and analyzed in our ISO/IEC 17025 accredited lab; traceable to at least two independent references (NIST, BAM or SI unit kg)