tri-Natriumcitrat Dihydrat ≥99,0%, Kristalle, AR® ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: Natriumcitrat tribasisch Dihydrat, Citronensäure Trinatriumsalz Dihydrat

0754-20EA 0 EUR
0754-20 MACR0754-06
tri-Natriumcitrat Dihydrat ≥99,0%, Kristalle, AR® ACS, Macron Fine Chemicals™
tri-Natriumcitrat Dihydrat
Formel: Na₃C₆H₅O₇·2H₂O
Molecular Weight: 294.1 g/mol
Schmelzpunkt: 150
Dichte: 1.70 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150031
CAS-Nummer: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HOC(COONa)(CH₂COONa)₂·2H₂O) ≥99.0 %
ACS - pH of 5% Solution at 25°C 7.0 - 9.0
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤0.003 %
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Ammonia (as NH₃) ≤0.003 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Iron (Fe) ≤5 ppm
Appearance (colorless or white crystals) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR