Essigsäure ≥99,0% ≥99,0%, TraceSELECT™ Ultra zur Ultraspurenanalyse, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Ethansäure

07692-250ML 07692-1L 07692-500ML
07692-250MLEA 435.2 EUR
07692-250ML 07692-1L 07692-500ML
Essigsäure ≥99,0% ≥99,0%, TraceSELECT™ Ultra zur Ultraspurenanalyse, Fluka™
Essigsäure

TraceSELECT™ Ultra and TraceSELECT


  • Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Formel: H₃CCOOH
Molecular Weight: 60.05 g/mol
Siedepunkt: 118 °C (1.013 hPa)
Schmelzpunkt: 16 °C
Dichte: 1.05 g/cm³ (20 °C)
Flammpunkt: 40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00036152
CAS-Nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 101.0 %
Silver (Ag) Max. 0.01 ppb
Aluminium (Al) Max. 0.2 ppb
Arsenic (As) Max. 0.5 ppb
Barium (Ba) Max. 0.01 ppb
Beryllium (Be) Max. 0.01 ppb
Bismuth (Bi) Max. 0.01 ppb
Calcium (Ca) Max. 1 ppb
Cadmium (Cd) Max. 0.01 ppb
Cerium (Ce) Max. 0.01 ppb
Cobalt (Co) Max. 0.01 ppb
Chromium (Cr) Max. 0.1 ppb
Cesium (Cs) Max. 0.01 ppb
Copper (Cu) Max. 0.2 ppb
Dysprosium (Dy) Max. 0.01 ppb
Erbium (Er) Max. 0.01 ppb
Europium (Eu) Max. 0.01 ppb
Iron (Fe) Max. 0.5 ppb
Gallium (Ga) Max. 0.01 ppb
Gadolinium (Gd) Max. 0.01 ppb
Germanium (Ge) Max. 0.02 ppb
Hafnium (Hf) Max. 0.01 ppb
Holmium (Ho) Max. 0.01 ppb
Indium (In) Max. 0.01 ppb
Potassium (K) Max. 0.5 ppb
Lanthanum (La) Max. 0.01 ppb
Lithium (Li) Max. 0.01 ppb
Lutetium (Lu) Max. 0.01 ppb
Magnesium (Mg) Max. 0.1 ppb
Manganese (Mn) Max. 0.02 ppb
Molybdenum (Mo) Max. 0.1 ppb
Sodium (Na) Max. 1 ppb
Neodymium (nd) Max. 0.01 ppb
Nickel (Ni) Max. 0.1 ppb
Lead (Pb) Max. 0.01 ppb
Praseodymium (Pr) Max. 0.01 ppb
Platinum (Pt) Max. 0.02 ppb
Rubidium (Rb) Max. 0.01 ppb
Rhenium (Re) Max. 0.01 ppb
Rhodium (Rh) Max. 0.01 ppb
Ruthenium (Ru) Max. 0.01 ppb
Antimony (Sb) Max. 0.05 ppb
Tin (Sn) Max. 1 ppb
Scandium (Sc) Max. 0.05 ppb
Selenium (Se) Max. 1 ppb
Samarium (Sm) Max. 0.01 ppb
Strontium (Sr) Max. 0.01 ppb
Terbium (Tb) Max. 0.01 ppb
Tellurium (Te) Max. 0.01 ppb
Thorium (Th) Max. 0.01 ppb
Titanium (Ti) Max. 0.05 ppb
Thallium (Tl) Max. 0.01 ppb
Thulium (Tm) Max. 0.01 ppb
Uran (U) Max. 0.01 ppb
Vanadium (V) Max. 0.01 ppb
Tungsten (W) Max. 0.01 ppb
Yttrium (Y) Max. 0.01 ppb
Ytterbium (Yb) Max. 0.01 ppb
Zinc (Zn) Max. 0.2 ppb
Zirconium (Zr) Max. 0.01 ppb
Autoignition Temperature 800 °F
Explosion Limit 4 %, 59 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR