Supelco® Organic Reference Standard, Octanoic Acid
Lieferant: Merck
07754-50MG
07754-50MGEA
86.4
EUR
07754-50MG
Supelco® Organic Reference Standard, Octanoic Acid
Standards
Organische Standards
- GHS signal: GHS05
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P273 - P280 - P303 + P361 + P353 - P304 + P340 + P310 - P305 + P351 + P338 - P363