Ammoniumdihydrogenphosphat ≥98,0%, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor

0776-01EA 102.2 EUR
0776-01 0776-07 0776-R
Ammoniumdihydrogenphosphat ≥98,0%, BAKER ANALYZED® ACS, J.T.Baker®
Ammoniumdihydrogenphosphat
Formel: NH₄H₂PO₄
Schmelzpunkt: 190 °C
Dichte: 1,80 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003396
CAS-Nummer: 7722-76-1
EINECS: 231-764-5
Merck Index: 13,00547

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements ≥98.0%
For Laboratory,Research,or Manufacturing Use ≤0.001%
Assay (NH₄H₂PO₄) ≥98.0 %
Calcium (Ca) ≤0.001 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 3.8 - 4.4
Magnesium (Mg) ≤0.0005 %
Nitrate (NO₃) ≤0.001 %
Sulfate (SO₄) ≤0.01 %
Trace Impurities - Iron (Fe) ≤0.001 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Chloride (Cl) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR