Supelco® Organic Reference Standard, 2,4-Dichlorophenol
Lieferant: Merck
07846-100MG
07846-100MGEA 117 EUR
07846-100MG
Supelco® Organic Reference Standard, 2,4-Dichlorophenol
Standards Organische Standards
  • GHS signal: GHS06, GHS05, GHS09
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P260 - P273 - P280 - P301 + P312 - P303 + P361 + P353 - P305 + P351 + P338
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55936411a0dbf2c2b46a6beaa6b96bafb8c8996c&vid=55936411a0dbf2c2b46a6beaa6b96bafb8c8996c&prid=web&clid=SAPDAM