Ammoniumsulfat 38% (w/w) in wässriger Lösung, BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor

Synonyme: Diammoniumsulfat, di-Ammoniumsulfat

0791-09EA 0 EUR
0791-09
Ammoniumsulfat 38% (w/w) in wässriger Lösung, BAKER ANALYZED®, J.T.Baker®
Ammoniumsulfat
Formel: (NH₄)₂SO₄
Molecular Weight: 132,14 g/mol
Dichte: 1,77 g/cm³
MDL: MFCD00003391
CAS-Nummer: 7783-20-2

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 37.0 - 39.0%
Assay ((NH₄)₂SO₄) (by formol method) 37.0 - 39.0 %
Color (APHA) ≤10
pH (As Is" basis)" 5.7 - 6.3
Identification A Passes Test
Identification B Passes Test
Insoluble Matter ≤0.005 %
Residue after Ignition ≤0.005 %
Nitrate (NO₃) ≤0.001 %
Chloride (Cl) ≤0.0005 %
Phosphate (PO₄) ≤0.0005 %
Arsenic (As) ≤0.00002 %
Heavy Metals (as Pb) ≤0.0005 %
Iron (Fe) ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR