Ammoniumsulfat 99,0-100,5%, körnig, BAKER ANALYZED® FCC, ACS für die Biochemie, für die Chromatographie Endotoxin getested, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Diammoniumsulfat, di-Ammoniumsulfat

0793-07EA 402.34 EUR
0793-07 0793-08 0793-09
Ammoniumsulfat 99,0-100,5%, körnig, BAKER ANALYZED® FCC, ACS für die Biochemie, für die Chromatographie Endotoxin getested, J.T.Baker®
Ammoniumsulfat

Ideal for biotechnology, cell biology and molecular biology discovery, and biopharmaceutical production, J.T.Baker® brand denaturants offer high purity, low insolubles and heavy metals with minimal contaminants. Extensive experience in managing the risks associated with handling and mixing large volumes of corrosive materials has enabled us to provide custom solutions perfectly suited to your needs.


  • Food GMP manufactured product

Formel: (NH₄)₂SO₄
Molecular Weight: 132.14 g/mol
Schmelzpunkt: 336…339 °C
Dichte: 1.78 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003391
CAS-Nummer: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Bestellen

Spezifikation Testergebnisse

Food GMP Manufactured Product 99.0 - 100.5%
Meets ACS Reagent Chemical Requirements ≤0.005%
Meets F.C.C. Requirements ≤0.005%
CAUTION: For Manufacturing, processing or repackaging 5.0 - 6.0
Bulk Food Chemical ≤0.001%
ACS - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
ACS - Insoluble Matter ≤0.005 %
ACS - Residue on Ignition (ACS) ≤0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 6.0
ACS - Nitrate (NO₃) ≤0.001 %
Chloride (Cl) ≤3 ppm
Phosphate (PO₄) ≤3 ppm
Heavy Metals (as Pb) ≤3 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
FCC - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
FCC - Identification Passes Test
FCC - Residue on Ignition (FCC) ≤0.25 %
FCC - Lead (Pb) ≤3 mg/kg
FCC - Selenium (Se) ≤0.003 %
Endotoxin Concentration (2.5 EU/g max.) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR