MPure-32™ aNAP Nucleic Acid Extraction System, MP Biomedicals
Lieferant: MP Biomedicals
07EMC043
07EMC043EA 19900 EUR
07EMC043
MPure-32™ aNAP Nucleic Acid Extraction System, MP Biomedicals
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
MPure-32™ aNAP System is a fully automated magnetic bead-based nucleic acid extraction system. It can process up to 32 samples simultaneously within a short period of time (around 40 to 60 minutes). Besides its capability to extract nucleic acid with high purity and yield, MPure-32™ aNAP System is easy to use; this eliminates the risk of human error and cross-contamination.
  • Superior Purity and Yield
  • High Flexibility (Open to Customisation)
  • Simple Workflow
  • Fast and Consistent Results
MP Biomedicals also offers a range of reagent kits (Magbeads FastDNA / FastRNA) for various types of biological samples. User has the flexibility to self-assemble the reagents with the relevant consumables for maximum cost-effectiveness or purchase the pre-assembled reagent kits for greater convenience.