Molekularsieb A4 (0,4 nm, 4 Å), BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor

Synonyme: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate

0810.1000EA 213.7 EUR
0810.1000
Molekularsieb A4 (0,4 nm, 4 Å), BAKER ANALYZED®, Laborreagens, J.T.Baker®
Molekularsieb A4 (0,4 nm, 4 Å)
Dichte: 1,1…2.2 g/cm³ (20 °C)
MDL: MFCD03457537
CAS-Nummer: 70955-01-0

Bestellen

Spezifikation Testergebnisse

Loss on Drying at 300°C max. 5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR