Salzsäure ≥30%, TraceSELECT™ Trace_analysis, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Hydrochloric acid solution, Muriatic acid

08256-100ML 08256-500ML 08256-1L 08256-2.5L
08256-100MLEA 105.56 EUR
08256-100ML 08256-500ML 08256-1L 08256-2.5L
Salzsäure ≥30%, TraceSELECT™ Trace_analysis, Fluka™
Salzsäure

TraceSELECT™ Ultra and TraceSELECT


  • Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Formel: HCl
Molecular Weight: 36.46 g/mol
Siedepunkt: 42 °C (1.013 hPa)
Schmelzpunkt: –35 °C
Dichte: 1.190 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Assay 30.0 - 32.0 %
Density (D 20/4) 1.140 - 1.160
Silver (Ag) Max. 0.5 ppb
Aluminium (Al) Max. 5 ppb
Arsenic (As) Max. 1 ppb
Gold (Au) Max. 1 ppb
Barium (Ba) Max. 0.5 ppb
Beryllium (Be) Max. 0.5 ppb
Bismuth (Bi) Max. 0.5 ppb
Calcium (Ca) Max. 10 ppb
Cadmium (Cd) Max. 0.5 ppb
Cobalt (Co) Max. 0.5 ppb
Chromium (Cr) Max. 1 ppb
Cesium (Cs) Max. 0.5 ppb
Copper (Cu) Max. 1 ppb
Iron (Fe) Max. 5 ppb
Gallium (Ga) Max. 0.5 ppb
Mercury (Hg) Max. 0.5 ppb
Indium (In) Max. 0.5 ppb
Iridium (Ir) Max. 0.5 ppb
Potassium (K) Max. 10 ppb
Lithium (Li) Max. 0.5 ppb
Magnesium (Mg) Max. 1 ppb
Manganese (Mn) Max. 0.5 ppb
Molybdenum (Mo) Max. 0.5 ppb
Sodium (Na) Max. 10 ppb
Nickel (Ni) Max. 1 ppb
Lead (Pb) Max. 0.5 ppb
Palladium (Pd) Max. 1 ppb
Platinum (Pt) Max. 0.5 ppb
Rubidium (Rb) Max. 0.5 ppb
Rhodium (Rh) Max. 0.5 ppb
Ruthenium (Ru) Max. 0.5 ppb
Antimony (Sb) Max. 0.5 ppb
Selenium (Se) Max. 0.001 ppm
Tin (Sn) Max. 1 ppb
Strontium (Sr) Max. 0.5 ppb
Titanium (Ti) Max. 1 ppb
Thallium (Tl) Max. 0.5 ppb
Uran (U) Max. 0.5 ppb
Vanadium (V) Max. 1 ppb
Zinc (Zn) Max. 1 ppb
Zirconium (Zr) Max. 0.5 ppb
Bromide (Br) Max. 10000 ppb
Free chlorine (Cl) Max. 0.1 ppm
Nitrate (NO₃) Max. 1000 ppb
Phosphate (PO₄) Max. 10 ppb
Sulfite (SO₃) Max. 1 ppm
Sulfate (SO₄) Max. 100 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR