Cetrimoniumbromid 99, high_purity
Lieferant: VWR Chemicals
|
Achtung
|
Synonyme:
Hexadecyltrimethylammoniumbromid, N-Cetyl-N,N,N-trimethylammoniumbromid, CTAB, N-Hexadecyl-N,N,N-trimethylammoniumbromid, Cetyltrimethylammoniumbromid
0833-1KGEA
636
EUR
0833-1KG
Cetrimoniumbromid 99, high_purity
Cetrimoniumbromid
A cationic positively-charged quaternary ammonium compounds. Utilised for applications ranging from solubilising membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis.
Formel:
CH₃(CH₂)₁₅N(Br)(CH₃)₃ Molecular Weight: 364.45 g/mol Schmelzpunkt: 237…243 °C Flammpunkt: 244 °C (geschlossener Tiegel) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011772 CAS-Nummer: 57-09-0 EINECS: 200-311-3 UN: 3077 ADR: 9 III Merck Index: 13,02034 |
Spezifikation Testergebnisse
Purity | ≥ 99 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...