Kieselgel, FluoroFlash® ~40 μm particle size, Supelco®

Lieferant: Merck

08965-100G-F
08965-100G-FEA 853 EUR
08965-100G-F
Kieselgel, FluoroFlash® ~40 μm particle size, Supelco®
Silica gel
Formel: SiO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 63231-67-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR