Knochenentkalker Q Path®, DC3, schnell wirkend
Lieferant: VWR Q-Path Chemicals
09128300.EA
96.9
EUR
09128300.
Knochenentkalker Q Path®, DC3, schnell wirkend
Färbemittel
Mikroskopie- und Färbereagenzien
Contains a mixture of hydrochloric acid and alcohols with some EDTA.
- Decalcify larger tissues (already fixated)
- Recommended volume is 20× the fragment volume
Ready to use for applications in histology and pathology laboratories.