Supelco® Organic Reference Standard, Benzbromarone
Lieferant: Merck
09147-50MG
09147-50MGEA 84.6 EUR
09147-50MG
Supelco® Organic Reference Standard, Benzbromarone
Standards Organische Standards
  • GHS signal: GHS06
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P301 + P330 + P331 + P310
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff4d535c81e1bc9659685781a32ee71a6db1abb3&vid=489530a41b5f553d012a367cf6f9121f70756ff8&prid=web&clid=SAPDAM