Organic reference standard, Pigment Orange 5, analytical standard, Supelco®
Lieferant: Merck
09215-25MG
09215-25MGEA 145 EUR
09215-25MG
Organic reference standard, Pigment Orange 5, analytical standard, Supelco®
Standards Organische Standards
Pigment orange 5 is an azo pigment, which can be produced industrially by a diazotisation and coupling sequence in which diazotised dinitroaniline is coupled into β-naphthol.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b4494dd7ae8079ead4306648d27858655604539&vid=8b4494dd7ae8079ead4306648d27858655604539&prid=web&clid=SAPDAM