Myco-Sniff-Valid™ Mycoplasma PCR Kit, MP Biomedicals
Lieferant: MP Biomedicals
093050301
093050301.EA
399
EUR
093050301.
Myco-Sniff-Valid™ Mycoplasma PCR Kit, MP Biomedicals
Tests
qPCR-Assays
Myco-Sniff-Valid™ mycoplasma PCR detection it provides a highly sensitive, specific and rapid method for the detection of a broad range of mycoplasma species in cell cultures.
- Easy to use: Contains premixed components for the PCR reaction, just add template DNA or samples
- Highly specific: No interference of animal or bacterial DNA
- Exogenous internal control: Helps differentiate false negatives due to PCR inhibition or erroneous PCR tests
The detection limit for contaminating mycoplasma is as low as 10 CFU/ml. The kit is sufficient for 48 tests (20 μl reactions).