L(+)-Ascorbinsäure 99,0-100,5%, kristallines Pulver USP, FCC, J.T.Baker®

Lieferant: Avantor

0936-07EA 255.83 EUR
0936-07
L(+)-Ascorbinsäure 99,0-100,5%, kristallines Pulver USP, FCC, J.T.Baker®
L(+)-Ascorbinsäure

  • Food GMP manufactured product

Formel: C₆H₈O₆
Siedepunkt: 553 °C (1013 hPa)
Schmelzpunkt: 190…192 °C
Dichte: 1,65 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064328
CAS-Nummer: 50-81-7
EINECS: 200-066-2
REACH: exempt
Merck Index: 13,00837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets F.C.C. Requirements 99.0 - 100.5%
Food GMP Manufactured Product Passes Test
GMP Manufactured Product Passes Test
Meets U.S.P Requirements 20.5 - 21.5°
CAUTION: For Manufacturing, processing or repackaging ≤0.1%
Bulk Food Chemical 99.0 - 100.5%
Bulk Pharmaceutical Chemical Passes Test
USP - Assay (C₆H₈O₆) 99.0 - 100.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Specific Rotation [Å]²⁵ ^D (+) 20.5 - 21.5 Degree
USP - Residue on Ignition ≤0.1 %
FCC - Assay (C₆H₈O₆) 99.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Specific Rotation [Å]²⁵ ^D (+) 20.5 - 21.5 Degree
FCC - Residue on Ignition ≤0.1 %
Arsenic (As) ≤3 ppm
Mesh - Thru U.S. No. 20 Sieve ≥100 %
Mesh - Thru U.S. No. 100 Sieve ≤70 %
Mesh - Thru U.S. No. 200 Sieve ≤20 %
Residual Solvents - Only Class 2 Solvents (Methanol) and Class 3
Solvents are likely to be present. Residual Class 2 Solvents are below
Option 1 and the residual concentration of all Class 3 Solvents is
<0.5%.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR