Span® 80, Supelco®

Lieferant: Merck

Synonyme: Arlacel 80, Sorbitan monooleate

M-Clarity™ Qualitätsstufe = MQ100

09569-50ML
09569-50MLEA 166 EUR
09569-50ML
Span® 80, Supelco®
Span® 80
Formel: C₂₄H₄₄O₆
Molecular Weight: 428,61 g/mol
Dichte: 1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00080948
CAS-Nummer: 1338-43-8
EINECS: 215-665-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e9947085272570ed7462805402d95d80a588a2&vid=c754ee438143ded2bcb1163b73622b88a93d2064&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR