L(+)-Ornithin Hydrochlorid ≥99%

Lieferant: VWR Chemicals

Synonyme: H-Orn-OH Hydrochloride, H-Orn-OH·HCl

0958-500GEA 463 EUR
0958-500G
L(+)-Ornithin Hydrochlorid ≥99%
L(+)-Ornithin Hydrochlorid
Formel: C₅H₁₂N₂O₂·HCl
Molecular Weight: 168,62 g/mol
MDL: MFCD00064562
CAS-Nummer: 3184-13-2
EINECS: 221-678-6

Bestellen

Spezifikation Testergebnisse

Ammonium = 0.1%
Heavy Metals = 10 ppm
Iron = 10 ppm
Loss on Drying = 0.5%
Purity = 99%
Specific Rotation (Dried, c=4, 6N HCl) 23 °- 25 °
Sulphate = 0.02%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR