Ammoniumformiat ≥97,0% (durch titrimetrische Analyse), Purum p.a.

Lieferant: Honeywell Chemicals
Achtung

09739-100G
09739-100GEA 28.52 EUR
09739-100G
Ammoniumformiat ≥97,0% (durch titrimetrische Analyse), Purum p.a.
Ammoniumformiat

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formel: HCO₂NH₄
Schmelzpunkt: 116 °C
Dichte: 1,266 g/cm3
Lagertemperatur: Raumtemperatur
MDL: MFCD00013103
CAS-Nummer: 540-69-2
EINECS: 208-753-9
Merck Index: 13,00525

Bestellen

Spezifikation Testergebnisse

Identity (IR) complying
Assay 97 - 103 %
pH (5 %, 25 °C) 6.0 - 7.0
Colour of the solution complying
Turbidity of solution complying
Water (Karl Fischer) Max. 3 %
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 200 ppm
Sodium (Na) Max. 100 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Chloride (Cl) Max. 50 ppm
Sulfate (SO₄) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR