Phenol ≥99,0% für die Biotechnologie pH 4.5

Lieferant: VWR Chemicals
Gefahr

0981-400MLEA 259 EUR
0981-400ML
Phenol ≥99,0% für die Biotechnologie pH 4.5
Phenol

An ideal replacement for water-saturated phenol in many protocols. Offers the high purity and performance of water saturated phenol with greatly enhanced stability and safety.


  • Manufactured using exceptionally high purity phenol that is double distilled & packaged under nitrogen
  • Liquid phenols offer a safer, easier alternative to crystalline phenol
  • Optimised pH for the purification of RNA
  • Long term stability > one year


For RNA purification, it is best to use a phenol or phenol:chloroform mixture with a pH of 4.5 to 5.5. Although phenol alone will purify RNA in these extractions, messenger RNA can collect with the DNA at the inter-phase in the complete absence of chloroform. The ratio of phenol to chloroform can vary from no chloroform to a 1:1 mixture, depending on the individual tissue and the character of the RNA being purified. For many applications, a 5:1 ratio of phenol:chloroform is a recommended starting point. The phenol used for extraction protocols must be highly purified and essentially free of contamination by phenolic acid, a product of phenol oxidation.

Formel: C₆H₅OH
Siedepunkt: 182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002143
CAS-Nummer: 108-95-2
EINECS: 203-632-7
UN: 2821
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR