di-Ammoniumhydrogencitrat ≥99,0% (durch titrimetrische Analyse), Puriss. p.a. ACS, Fluka™

Lieferant: Honeywell Chemicals
Achtung

Synonyme: sek-Ammoniumcitrat, Diammoniumhydrogen-2-hydroxypropan-1,2,3-tricarboxylat, Ammoniumcitrat dibasisch, Citronensäure Diammoniumsalz

09831-500G
09831-500GEA 91.07 EUR
09831-500G
di-Ammoniumhydrogencitrat ≥99,0% (durch titrimetrische Analyse), Puriss. p.a. ACS, Fluka™
di-Ammoniumhydrogencitrat
Formel: HOC(CO₂H)(CH₂CO₂NH₄)₂
Molecular Weight: 226.19 g/mol
Dichte: 1.480 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013068
CAS-Nummer: 3012-65-5
EINECS: 221-146-3
Merck Index: 13,00513

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 101.0 %
Residue on ignition Max. 0.01 %
Water insoluble matter Max. 0.005 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 500 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 5 ppm
Oxalate (C₂O₄) Max. 500 ppm
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR