Nonactin 95,0 HPLC_ASSAY_METHOD, Selectophore™, Funtion tested ionophore, Supelco®
Lieferant: Merck
Synonyme:
Polynactin, Werramycin A
MQ100
09877-250MG
09877-50MG
09877-250MGEA
1620
EUR
09877-250MG
09877-50MG
Nonactin 95,0 HPLC_ASSAY_METHOD, Selectophore™, Funtion tested ionophore, Supelco®
Nonactin
Transports alkali cations through biological membranes, with high selectivity for ammonia.
Determination of potassium ions in pharmaceutical samples by FIA using a potentiometric electrode based on ionophore nonactin occluded in EVA membrane.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel:
C₄₀H₆₄O₁₂ Molecular Weight: 736.9 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
6833-84-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...