Nonactin 95,0 HPLC_ASSAY_METHOD, Selectophore™, Funtion tested ionophore, Supelco®

Lieferant: Merck

Synonyme: Polynactin, Werramycin A

MQ100

09877-250MG 09877-50MG
09877-250MGEA 1620 EUR
09877-250MG 09877-50MG
Nonactin 95,0 HPLC_ASSAY_METHOD, Selectophore™, Funtion tested ionophore, Supelco®
Nonactin

Transports alkali cations through biological membranes, with high selectivity for ammonia.


Determination of potassium ions in pharmaceutical samples by FIA using a potentiometric electrode based on ionophore nonactin occluded in EVA membrane.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₄₀H₆₄O₁₂
Molecular Weight: 736.9 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 6833-84-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb6d4c00294e2f6425452e22d4b107b9daec7fff&vid=95d475f480b5b3ae67333d130cf23a65aec75ca0&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR