Ammoniumheptamolybdat Tetrahydrat ≥99,0% (durch titrimetrische Analyse), Puriss. p.a. ACS, Fluka™
Lieferant: Honeywell Chemicals
09880-1KG
09880-100G
09880-500G
09880-25KG
09880-1KGEA
593.08
EUR
09880-1KG
09880-100G
09880-500G
09880-25KG
Ammoniumheptamolybdat Tetrahydrat ≥99,0% (durch titrimetrische Analyse), Puriss. p.a. ACS, Fluka™
Ammoniumheptamolybdat Tetrahydrat
Formel:
(NH₄)₆Mo₇O₂₄·4H₂O Dichte: 2,498 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00167059 CAS-Nummer: 12054-85-2 Merck Index: 13,00536 |
Spezifikation Testergebnisse
Assay | 99.4 - 101.8 % |
Assay (as MoO₃) | 81.0 - 83.0 % |
Water insoluble matter | Max. 50 ppm |
Calcium (Ca) | Max. 10 ppm |
Cadmium (Cd) | Max. 5 ppm |
Cobalt (Co) | Max. 5 ppm |
Chromium (Cr) | Max. 5 ppm |
Copper (Cu) | Max. 5 ppm |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 50 ppm |
Magnesium (Mg) | Max. 5 ppm |
Manganese (Mn) | Max. 5 ppm |
Sodium (Na) | Max. 50 ppm |
Nickel (Ni) | Max. 5 ppm |
Lead (Pb) | Max. 5 ppm |
Zinc (Zn) | Max. 5 ppm |
Heavy metals (as Pb) | Max. 10 ppm |
Heavy metals (ICP) ACS | Max. 0.001 % |
Chloride (Cl) | Max. 10 ppm |
Nitrate (NO₃) | Max. 20 ppm |
Phosphate (PO₄) | Max. 5 ppm |
Phosphate, Arsenate, Silicate (as SiO₂) | Max. 10 ppm |
Sulfate (SO₄) | Max. 100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...