Ammoniumheptamolybdat Tetrahydrat ≥99,0% (durch titrimetrische Analyse), Puriss. p.a. ACS, Fluka™

Lieferant: Honeywell Chemicals

09880-1KG 09880-100G 09880-500G 09880-25KG
09880-1KGEA 593.08 EUR
09880-1KG 09880-100G 09880-500G 09880-25KG
Ammoniumheptamolybdat Tetrahydrat ≥99,0% (durch titrimetrische Analyse), Puriss. p.a. ACS, Fluka™
Ammoniumheptamolybdat Tetrahydrat
Formel: (NH₄)₆Mo₇O₂₄·4H₂O
Dichte: 2,498 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00167059
CAS-Nummer: 12054-85-2
Merck Index: 13,00536

Bestellen

Spezifikation Testergebnisse

Assay 99.4 - 101.8 %
Assay (as MoO₃) 81.0 - 83.0 %
Water insoluble matter Max. 50 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 10 ppm
Heavy metals (ICP) ACS Max. 0.001 %
Chloride (Cl) Max. 10 ppm
Nitrate (NO₃) Max. 20 ppm
Phosphate (PO₄) Max. 5 ppm
Phosphate, Arsenate, Silicate (as SiO₂) Max. 10 ppm
Sulfate (SO₄) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR