Acetonitril, EMSURE® Reag. Ph. Eur., ACS zur Analyse, Supelco®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Ethannitril, Essigsäurenitril, Ethansäurenitril, Methylcyanid
M-Clarity™ Qualitätsstufe = MQ300
1.00003.1000
1.00003.2500
1.00003.6010
1.00003.6025
1.00003.9950
1.00003.4000
1.00003.1000EA
124
EUR
1.00003.1000
1.00003.2500
1.00003.6010
1.00003.6025
1.00003.9950
1.00003.4000
Acetonitril, EMSURE® Reag. Ph. Eur., ACS zur Analyse, Supelco®
Acetonitril
EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. Merck declare the EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
Formel:
H₃CCN Molecular Weight: 41.05 g/mol Siedepunkt: 81.6 Schmelzpunkt: –45.7 °C Dichte: 0.782 g/cm³ (20 °C) Flammpunkt: 2 °C (geschlossener Tiegel) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001878 CAS-Nummer: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3 II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Spezifikation Testergebnisse
Solubility | (20 °C) soluble |
Melting Point | -45.7 °C |
Molar Mass | 41.05 g/mol |
Boiling Point | 81.6 °C (1013 hPa) |
Vapor Pressure | 97 hPa (20 °C) |
Flash Point | 2 °C |
Refractive Index | 1.34 |
Explosion Limit | 3.0 - 17 %(V) |
Density | 0.786 g/cm3 (20 °C) |
Ignition Point | 524 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...