Aceton, EMSURE® ACS, ISO, Reag. Ph. Eur. zur Analyse, Supelco®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ300
EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. Merck declares their EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph. Eur.) and also with the ISO standards.
Formel:
CH₃COCH₃ Siedepunkt: 56,2 °C (1013 hPa) Dichte: 0,792 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008765 CAS-Nummer: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II REACH: 01-2119471330-49 Merck Index: 13,00067 |
|
Spezifikation Testergebnisse
Solubility | (20 °C) soluble |
Melting Point | -95.4 °C |
Molar Mass | 58.08 g/mol |
Boiling Point | 56.2 °C (1013 hPa) |
Vapor Pressure | 233 hPa (20 °C) |
Refractive Index | 1.35868 (20 °C) |
Explosion Limit | 2.6 - 12.8%(V) |
Density | 0.79 g/cm³ (20 °C) |
pH | 5 - 6 (395 g/L, H₂O, 20 °C) |
Ignition Point | 465 °C DIN 51794 |
Water Absorption | 1000 g/kg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...