Essigsäure glacial_AcOH 99,8-100,5 Alkalimetric, EMPROVE® ESSENTIAL USP, JP, E260, Ph. Eur., BP suitable for use as excipient, SAFC®

Lieferant: Merck
Gefahr

Synonyme: Ethansäure

1.00056.2500 1.00056.9190 1.00056.9025 1.00056.5000 1.00056.9960 1.00056.9950
1.00056.2500EA 300 EUR
1.00056.2500 1.00056.9190 1.00056.9025 1.00056.5000 1.00056.9960 1.00056.9950
Essigsäure glacial_AcOH 99,8-100,5 Alkalimetric, EMPROVE® ESSENTIAL USP, JP, E260, Ph. Eur., BP suitable for use as excipient, SAFC®
Essigsäure

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Standards: Corresponds to Ph. Eur., BP, JP, USP, E260. Conforms to the purity criteria on food additives according to the current European Commission Regulation.

Formel: H₃CCOOH
Molecular Weight: 60.05 g/mol
Siedepunkt: 118
Schmelzpunkt: 17
Dichte: 1.05 g/cm³ (20 °C)
Flammpunkt: 38,5
Lagertemperatur: Raumtemperatur
MDL: MFCD00036152
CAS-Nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3) II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (alkalimetric) 99,8 - 100,5 %
Identity passes test
Appearance of the substance passes test
Solidification temperature ≥ 15,6 °C
Foreign organic substances passes test
Formic acid, Formates and other oxidable impurities (as HCOOH) ≤ 0,10 %
Heavy metals (as Pb) ≤ 0,0005 %
Chloride (Cl) ≤ 0,0002 %
Sulfate (SO₄) ≤ 0,0005 %
Al (Aluminium) ≤ 0,00005 %
As (Arsenic) ≤ 0,00010 %
Cu (Copper) ≤ 0,0005 %
Fe (Iron) ≤ 0,0005 %
Hg (Mercury) ≤ 0,00010 %
Pb (Lead) ≤ 0,00005 %
Zn (Zinc) ≤ 0,0005 %
Other residual solvents (ICH Q3C) ...excluded by production process
Reducing substances (Ph Eur) passes test
Substances reducing potassium permanganate passes test
Evaporation residue ≤ 0,005 %
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph Eur, BP, JP, USP, E260
Conforms to the purity criteria on food additives according to the current European Commission Regulation.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR