Essigsäure (Eisessig), wasserfrei 100% (durch Alkalimetrie), EMSURE® ACS, ISO, Reag. Ph. Eur. zur Analyse, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ300

1.00063.1000 1.00063.1011 1.00063.2500 1.00063.2511 1.00063.9026 1.00063.9200 1.00063.2510 1.00063.0510 1.00063.1010
1.00063.1000EA 71.4 EUR
1.00063.1000 1.00063.1011 1.00063.2500 1.00063.2511 1.00063.9026 1.00063.9200 1.00063.2510 1.00063.0510 1.00063.1010
Essigsäure (Eisessig), wasserfrei 100% (durch Alkalimetrie), EMSURE® ACS, ISO, Reag. Ph. Eur. zur Analyse, Supelco®
Essigsäure
Formel: H₃CCOOH
Siedepunkt: 118 °C (1013 hPa)
Schmelzpunkt: 17 °C
Dichte: 1,05 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00036152
CAS-Nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3),II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (Alkalimetric) ≥99.8%
Identity Passes Test
Color ≤10 Hazen
Acetaldehyde ≤2 ppm
Acetic Anhydride ≤100 ppm
Titratable Base ≤0.0004 meq/g
Solidification Temperature ≥16.3 °C
Chloride (Cl) ≤0.4 ppm
Phosphate (PO₄) ≤0.4 ppm
Heavy metals (as Pb) ≤0.5 ppm
Sulfate (SO₄) ≤0.4 ppm
Ag (Silver) ≤0.005 ppm
Al (Aluminium) ≤0.020 ppm
As (Arsenic) ≤0.010 ppm
Au (Gold) ≤0.010 ppm
B (Boron) ≤0.100 ppm
Ba (Barium) ≤0.010 ppm
Be (Beryllium) ≤0.005 ppm
Bi (Bismuth) ≤0.050 ppm
Ca (Calcium) ≤0.100 ppm
Cd (Cadmium) ≤0.020 ppm
Co (Cobalt) ≤0.010 ppm
Cr (Chromium) ≤0.020 ppm
Cu (Copper) ≤0.010 ppm
Fe (Iron) ≤0.050 ppm
Ga (Gallium) ≤0.050 ppm
Ge (Germanium) ≤0.020 ppm
Hg (Mercury) ≤0.005 ppm
In (Indium) ≤0.050 ppm
K (Potassium) ≤0.100 ppm
Li (Lithium) ≤0.010 ppm
Mg (Magnesium) ≤0.050 ppm
Mn (Manganese) ≤0.010 ppm
Mo (Molybdenum) ≤0.010 ppm
Na (Sodium) ≤0.200 ppm
Ni (Nickel) ≤0.020 ppm
Pb (Lead) ≤0.010 ppm
Pt (Platinum) ≤0.100 ppm
Sn (Tin) ≤0.050 ppm
Sr (Strontium) ≤0.010 ppm
Ti (Titanium) ≤0.050 ppm
Tl (Thallium) ≤0.020 ppm
V (Vanadium) ≤0.010 ppm
Zn (Zinc) ≤0.030 ppm
Zr (Zirconium) ≤0.050 ppm
Substances Reducing Potassium Dichromate Passes Test
Substances Reducing Potassium Permanganate ≤20 ppm
Evaporation Residue ≤5 ppm
Water ≤0.2
Dilution test Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR