Essigsäure (Eisessig), wasserfrei 100% (durch Alkalimetrie), EMSURE® ACS, ISO, Reag. Ph. Eur. zur Analyse, Supelco®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ300
1.00063.1000
1.00063.1011
1.00063.2500
1.00063.2511
1.00063.9026
1.00063.9200
1.00063.2510
1.00063.0510
1.00063.1010
1.00063.1000EA
71.4
EUR
1.00063.1000
1.00063.1011
1.00063.2500
1.00063.2511
1.00063.9026
1.00063.9200
1.00063.2510
1.00063.0510
1.00063.1010
Essigsäure (Eisessig), wasserfrei 100% (durch Alkalimetrie), EMSURE® ACS, ISO, Reag. Ph. Eur. zur Analyse, Supelco®
Essigsäure
Formel:
H₃CCOOH Siedepunkt: 118 °C (1013 hPa) Schmelzpunkt: 17 °C Dichte: 1,05 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036152 CAS-Nummer: 64-19-7 EINECS: 200-580-7 UN: 2789 ADR: 8 (3),II REACH: 01-2119475328-30 |
|
Spezifikation Testergebnisse
Assay (Alkalimetric) | ≥99.8% |
Identity | Passes Test |
Color | ≤10 Hazen |
Acetaldehyde | ≤2 ppm |
Acetic Anhydride | ≤100 ppm |
Titratable Base | ≤0.0004 meq/g |
Solidification Temperature | ≥16.3 °C |
Chloride (Cl) | ≤0.4 ppm |
Phosphate (PO₄) | ≤0.4 ppm |
Heavy metals (as Pb) | ≤0.5 ppm |
Sulfate (SO₄) | ≤0.4 ppm |
Ag (Silver) | ≤0.005 ppm |
Al (Aluminium) | ≤0.020 ppm |
As (Arsenic) | ≤0.010 ppm |
Au (Gold) | ≤0.010 ppm |
B (Boron) | ≤0.100 ppm |
Ba (Barium) | ≤0.010 ppm |
Be (Beryllium) | ≤0.005 ppm |
Bi (Bismuth) | ≤0.050 ppm |
Ca (Calcium) | ≤0.100 ppm |
Cd (Cadmium) | ≤0.020 ppm |
Co (Cobalt) | ≤0.010 ppm |
Cr (Chromium) | ≤0.020 ppm |
Cu (Copper) | ≤0.010 ppm |
Fe (Iron) | ≤0.050 ppm |
Ga (Gallium) | ≤0.050 ppm |
Ge (Germanium) | ≤0.020 ppm |
Hg (Mercury) | ≤0.005 ppm |
In (Indium) | ≤0.050 ppm |
K (Potassium) | ≤0.100 ppm |
Li (Lithium) | ≤0.010 ppm |
Mg (Magnesium) | ≤0.050 ppm |
Mn (Manganese) | ≤0.010 ppm |
Mo (Molybdenum) | ≤0.010 ppm |
Na (Sodium) | ≤0.200 ppm |
Ni (Nickel) | ≤0.020 ppm |
Pb (Lead) | ≤0.010 ppm |
Pt (Platinum) | ≤0.100 ppm |
Sn (Tin) | ≤0.050 ppm |
Sr (Strontium) | ≤0.010 ppm |
Ti (Titanium) | ≤0.050 ppm |
Tl (Thallium) | ≤0.020 ppm |
V (Vanadium) | ≤0.010 ppm |
Zn (Zinc) | ≤0.030 ppm |
Zr (Zirconium) | ≤0.050 ppm |
Substances Reducing Potassium Dichromate | Passes Test |
Substances Reducing Potassium Permanganate | ≤20 ppm |
Evaporation Residue | ≤5 ppm |
Water | ≤0.2 |
Dilution test | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...