Nuclear fast red-aluminum sulphate solution 0,1% Verwendung in der nuklearen Färbung, für die Histologie, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
For nuclear staining and for counterstaining in histochemical reactions e.g. Berlin blue
Nuclear fast red-aluminum sulfate solution 0.1% is used for counterstaining in histochemical reactions, e.g. in Berlin blue staining for detection of iron or in silver staining of reticulin fibers. Nuclear fast red-aluminum sulfate solution 0.1% can also be used for nuclear staining with Alcian blue. 500 ml of the Nuclear fast red-aluminum sulfate solution 0.1% are sufficient for staining at least 500 specimens. The solution has an excellent quality as it is an IVD registered product and is CE certified, thus, it can be used in an accredited laboratory.
![]() |
Spezifikation Testergebnisse
Suitability for microscopy | passes test |
Nuclei | dark red |
Cytoplasm | light red |
Acid mucosubstances | light blue |